COM945512528 Combi-Blocks, Inc.

1-(2-Aminoethyl)-1h-pyrrole-2,5-dione

  • MDL number MFCD14619234
  • Cas Number : 125923-10-6
  • Molecular Weight : 140.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-4052

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-4052
Availability details for
Quantity
  • Availability
  • Add to Cart