COM945512545 Combi-Blocks, Inc.

2-Bromothiazol-4-amine

  • MDL number MFCD11040376
  • Cas Number : 41731-33-3
  • Molecular Weight : 179.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-1454

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-1454
Availability details for
Quantity
  • Availability
  • Add to Cart