COM945512546 Combi-Blocks, Inc.

Pyridine-2-carboxamidine

  • MDL number MFCD05663356
  • Cas Number : 52313-50-5
  • Molecular Weight : 121.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-1683

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-1683
Availability details for
Quantity
  • Availability
  • Add to Cart