COM945512642 Combi-Blocks, Inc.

1-Methyl-6-oxopyridine-3-carbaldehyde

  • MDL number MFCD24674768
  • Cas Number : 98279-50-6
  • Molecular Weight : 137.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2336

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2336
Availability details for
Quantity
  • Availability
  • Add to Cart