COM945512729 Combi-Blocks, Inc.

1-(2,3-Dihydro-1,4-benzodioxin-6-yl)methanamine, HCl

  • MDL number MFCD08445280
  • Cas Number : 31127-40-9
  • Molecular Weight : 201.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8269

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8269
Availability details for
Quantity
  • Availability
  • Add to Cart