COM964462724 Combi-Blocks, Inc.

4-Bromo-2-nitrophenoxyacetic acid

  • MDL number MFCD06823836
  • Cas Number : 128627-49-6
  • Molecular Weight : 276.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6002

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6002
Availability details for
Quantity
  • Availability
  • Add to Cart