COM964462732 Combi-Blocks, Inc.

N-Acetyl-N-(2,6-dimethylphenyl)acetamide

  • MDL number MFCD30834331
  • Cas Number : 91573-23-8
  • Molecular Weight : 205.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6249

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6249
Availability details for
Quantity
  • Availability
  • Add to Cart