COM964462738 Combi-Blocks, Inc.

[2-Hydroxy-3-(methoxycarbonyl)phenyl]boronic acid

  • MDL number MFCD30834335
  • Cas Number : 2304635-49-0
  • Molecular Weight : 195.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4521

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4521
Availability details for
Quantity
  • Availability
  • Add to Cart