COM964462758 Combi-Blocks, Inc.

(2S)-1-[(4-Bromophenyl)carbonyl]-2-methylpyrrolidine-2-carboxylic acid

  • MDL number MFCD30834351
  • Cas Number : 2140265-41-2
  • Molecular Weight : 312.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5971

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5971
Availability details for
Quantity
  • Availability
  • Add to Cart