COM964462781 Combi-Blocks, Inc.

[4-({1-[(tert-butoxy)carbonyl]piperidin-4-yl}oxy)-2,6-difluorophenyl]boronic acid

  • MDL number MFCD30834359
  • Cas Number : 2377606-84-1
  • Molecular Weight : 357.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4441

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4441
Availability details for
Quantity
  • Availability
  • Add to Cart