COM964462794 Combi-Blocks, Inc.

t-Butyl Z-L-Alaninamide

  • MDL number MFCD30834371
  • Cas Number : 871579-11-2
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-0907

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-0907
Availability details for
Quantity
  • Availability
  • Add to Cart