COM964462795 Combi-Blocks, Inc.

4-Amino-2-chloro-N-(isobutyl)benzamide

  • MDL number MFCD09044583
  • Cas Number : 926218-95-3
  • Molecular Weight : 226.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1649

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1649
Availability details for
Quantity
  • Availability
  • Add to Cart