COM964462802 Combi-Blocks, Inc.

2-Chloro-6-nitropyridin-3-ol

  • MDL number MFCD28668568
  • Cas Number : 15206-27-6
  • Molecular Weight : 174.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1896

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1896
Availability details for
Quantity
  • Availability
  • Add to Cart