COM964462830 Combi-Blocks, Inc.

2-Methylthio-3,5-methylpyrazine

  • MDL number MFCD00016740
  • Cas Number : 67952-65-2
  • Molecular Weight : 154.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?NA-0277

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?NA-0277
Availability details for
Quantity
  • Availability
  • Add to Cart