COM964462832 Combi-Blocks, Inc.

Boc-met-gly-oh

  • MDL number MFCD00155462
  • Cas Number : 23446-03-9
  • Molecular Weight : 306.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?NA-0281

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?NA-0281
Availability details for
Quantity
  • Availability
  • Add to Cart