COM964462833 Combi-Blocks, Inc.

L-Cysteinylglycine monohydrate

  • MDL number MFCD00754496
  • Cas Number : 19246-18-5
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 85
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?NA-0283

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?NA-0283
Availability details for
Quantity
  • Availability
  • Add to Cart