COM964462875 Combi-Blocks, Inc.

1-Benzylpyrrolidine-2-carboxylic acid hydrochloride

  • MDL number MFCD00157011
  • Cas Number : 92086-93-6
  • Molecular Weight : 241.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4360

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4360
Availability details for
Quantity
  • Availability
  • Add to Cart