COM964462887 Combi-Blocks, Inc.

Methyl 6-bromo-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline-8-carboxylate

  • MDL number MFCD30834417
  • Cas Number : 2377606-22-7
  • Molecular Weight : 392.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1409

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1409
Availability details for
Quantity
  • Availability
  • Add to Cart