COM964462896 Combi-Blocks, Inc.

5-Fluoro-2-isopropoxyphenylboronic acid pinacol ester

  • MDL number MFCD29092285
  • Cas Number : 1497435-85-4
  • Molecular Weight : 280.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8193

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8193
Availability details for
Quantity
  • Availability
  • Add to Cart