COM964462926 Combi-Blocks, Inc.

3-Heptyne

  • MDL number MFCD00039960
  • Cas Number : 2586-89-2
  • Molecular Weight : 96.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4781

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4781
Availability details for
Quantity
  • Availability
  • Add to Cart