COM964462964 Combi-Blocks, Inc.

1-(2-Chloroethyl)-4-methoxybenzene

  • MDL number MFCD00044718
  • Cas Number : 18217-00-0
  • Molecular Weight : 170.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5875

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5875
Availability details for
Quantity
  • Availability
  • Add to Cart