COM964463042 Combi-Blocks, Inc.

Bis-peg13-t-butyl ester

  • MDL number MFCD30182036
  • Cas Number : 2225903-65-9
  • Molecular Weight : 803

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0017

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0017
Availability details for
Quantity
  • Availability
  • Add to Cart