COM964463113 Combi-Blocks, Inc.

P-Tolyl isobutyrate

  • MDL number MFCD00026440
  • Cas Number : 103-93-5
  • Molecular Weight : 178.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0232

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0232
Availability details for
Quantity
  • Availability
  • Add to Cart