COM964463138 Combi-Blocks, Inc.

(S)-2,3,10,11A-Tetrahydro-1h-benzo[e]pyrrolo[1,2-a][1,4]diazepin-11(5h)-one

  • MDL number MFCD30471427
  • Cas Number : 59824-68-9
  • Molecular Weight : 202.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0308

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0308
Availability details for
Quantity
  • Availability
  • Add to Cart