COM964463197 Combi-Blocks, Inc.

(E)-N-(2-Fluorobenzylidene)-2-methylpropan-2-amine

  • MDL number MFCD27946857
  • Cas Number : 195004-63-8
  • Molecular Weight : 179.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0466

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0466
Availability details for
Quantity
  • Availability
  • Add to Cart