COM964463225 Combi-Blocks, Inc.

4-[(4-Bromophenyl)(4-methylphenyl)amino]benzaldehyde

  • MDL number MFCD16658914
  • Cas Number : 733744-98-4
  • Molecular Weight : 366.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0538

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0538
Availability details for
Quantity
  • Availability
  • Add to Cart