COM964463258 Combi-Blocks, Inc.

2-(7-Methyl-1,2-benzoxazol-3-yl)acetic acid

  • MDL number MFCD05883978
  • Cas Number : 34172-93-5
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0657

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0657
Availability details for
Quantity
  • Availability
  • Add to Cart