COM964463271 Combi-Blocks, Inc.

N2,N6-Bis(2-chloroethyl)pyridine-2,6-dicarboxamide

  • MDL number MFCD28347675
  • Cas Number : 345617-16-5
  • Molecular Weight : 290.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0700

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0700
Availability details for
Quantity
  • Availability
  • Add to Cart