COM964463286 Combi-Blocks, Inc.

Ethyl 2-(4-(nitromethyl)tetrahydro-2h-pyran-4-yl)acetate

  • MDL number MFCD29917052
  • Cas Number : 205827-21-0
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0734

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0734
Availability details for
Quantity
  • Availability
  • Add to Cart