COM964463292 Combi-Blocks, Inc.

Methyl 2-amino-4-chloroquinazoline-7-carboxylate

  • MDL number MFCD30468801
  • Cas Number : 1246209-90-4
  • Molecular Weight : 237.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0753

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0753
Availability details for
Quantity
  • Availability
  • Add to Cart