COM964463349 Combi-Blocks, Inc.

3-(tert-Butyl)-6-chloro-1,6-dihydropyridazine

  • MDL number MFCD29921157
  • Cas Number : 1956331-91-1
  • Molecular Weight : 172.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0908

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0908
Availability details for
Quantity
  • Availability
  • Add to Cart