COM964463371 Combi-Blocks, Inc.

1,3-Di-tert-butyl 2-oxopyrrolidine-1,3-dicarboxylate

  • MDL number MFCD24471094
  • Cas Number : 1180519-44-1
  • Molecular Weight : 285.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0978

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0978
Availability details for
Quantity
  • Availability
  • Add to Cart