COM964463391 Combi-Blocks, Inc.

2-(2-(2-(Docosyloxy)ethoxy)ethoxy) ethanol

  • MDL number MFCD29090723
  • Cas Number : 297176-96-6
  • Molecular Weight : 458.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1033

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1033
Availability details for
Quantity
  • Availability
  • Add to Cart