COM964463415 Combi-Blocks, Inc.

3-(Benzyloxy)-1-(2,3-dihydroxypropyl)-4-oxo-1,4-dihydropyridine-2-carboxylic acid

  • MDL number MFCD22741606
  • Cas Number : 1206102-06-8
  • Molecular Weight : 319.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1121

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1121
Availability details for
Quantity
  • Availability
  • Add to Cart