COM964463445 Combi-Blocks, Inc.

1-Bromo-3-fluoro-5-vinylbenzene

  • MDL number MFCD27926579
  • Cas Number : 627527-35-9
  • Molecular Weight : 201.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1200

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1200
Availability details for
Quantity
  • Availability
  • Add to Cart