COM964463446 Combi-Blocks, Inc.

(S)-Methyl 2-amino-2-(4-(benzyloxy)phenyl)acetate

  • MDL number MFCD22677490
  • Cas Number : 908066-52-4
  • Molecular Weight : 271.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1202

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1202
Availability details for
Quantity
  • Availability
  • Add to Cart