COM964463466 Combi-Blocks, Inc.

2-Chloro-n-(2-chloro-4-methylpyridin-3-yl)nicotinamide

  • MDL number MFCD09031256
  • Cas Number : 133627-46-0
  • Molecular Weight : 282.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1264

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1264
Availability details for
Quantity
  • Availability
  • Add to Cart