COM964463530 Combi-Blocks, Inc.

Dipyrido[3,2-a:2',3'-c]phenazine

  • MDL number MFCD28125537
  • Cas Number : 19535-47-8
  • Molecular Weight : 282.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1468

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1468
Availability details for
Quantity
  • Availability
  • Add to Cart