COM964463542 Combi-Blocks, Inc.

2-((2-Oxo-2h-chromen-7-yl)oxy)-n-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide

  • MDL number MFCD29090683
  • Molecular Weight : 421.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1497

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1497
Availability details for
Quantity
  • Availability
  • Add to Cart