COM964463550 Combi-Blocks, Inc.

4-Anilino-1,1':4',1''-terphenyl

  • MDL number MFCD28963769
  • Cas Number : 897671-81-7
  • Molecular Weight : 321.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1514

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1514
Availability details for
Quantity
  • Availability
  • Add to Cart