COM964463555 Combi-Blocks, Inc.

2-(9,9-Dimethyl-9h-fluoren-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD30187267
  • Cas Number : 1365692-79-0
  • Molecular Weight : 320.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1527

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1527
Availability details for
Quantity
  • Availability
  • Add to Cart