COM964463558 Combi-Blocks, Inc.

2-(3-(Chloromethyl)phenyl)-2h-1,2,3-triazole

  • MDL number MFCD29921021
  • Cas Number : 1956328-00-9
  • Molecular Weight : 193.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1533

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1533
Availability details for
Quantity
  • Availability
  • Add to Cart