COM964463633 Combi-Blocks, Inc.

(S)-tert-Butyl 2-(aminomethyl)pyrrolidine-1-carboxylate hydrochloride

  • MDL number MFCD11101392
  • Cas Number : 1190890-11-9
  • Molecular Weight : 236.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1750

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1750
Availability details for
Quantity
  • Availability
  • Add to Cart