COM964463637 Combi-Blocks, Inc.

tert-Butyl 3,3-dimethyl-2-oxopyrrolidine-1-carboxylate

  • MDL number MFCD15071758
  • Cas Number : 153039-16-8
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1760

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1760
Availability details for
Quantity
  • Availability
  • Add to Cart