COM964463649 Combi-Blocks, Inc.

tert-Butyl 4-(2-methylpyrimidin-5-yl)piperazine-1-carboxylate

  • MDL number MFCD29921321
  • Cas Number : 1956380-47-4
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1789

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1789
Availability details for
Quantity
  • Availability
  • Add to Cart