COM964463681 Combi-Blocks, Inc.

1-(2,3-Dimethylphenyl)-1-(1h-imidazol-5-yl)ethanol

  • MDL number MFCD29090692
  • Cas Number : 86347-12-8
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1888

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1888
Availability details for
Quantity
  • Availability
  • Add to Cart