COM964463684 Combi-Blocks, Inc.

6-(Trifluoromethyl)indole-3-carboxaldehyde

  • MDL number MFCD09954784
  • Cas Number : 13544-09-7
  • Molecular Weight : 213.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1899

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1899
Availability details for
Quantity
  • Availability
  • Add to Cart