COM964463700 Combi-Blocks, Inc.

2-Amino-4-(trifluoromethyl)benzaldehyde

  • MDL number MFCD10696884
  • Cas Number : 109466-88-8
  • Molecular Weight : 189.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1940

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1940
Availability details for
Quantity
  • Availability
  • Add to Cart