COM964463714 Combi-Blocks, Inc.

(3R,4S)-Methyl 1-benzyl-4-p-tolylpyrrolidine-3-carboxylate

  • MDL number MFCD20489350
  • Cas Number : 898547-79-0
  • Molecular Weight : 309.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1983

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1983
Availability details for
Quantity
  • Availability
  • Add to Cart