COM964463811 Combi-Blocks, Inc.

2-Chloro-8-methoxyquinazolin-4-amine

  • MDL number MFCD11858270
  • Cas Number : 61948-65-0
  • Molecular Weight : 209.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2242

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2242
Availability details for
Quantity
  • Availability
  • Add to Cart